Learn More
Waters Corp Preparative Column, HSS T3, 100Å, 5 μm, 10 mm x 250 mm
Supplier:  Waters Corp 186008280
Encompass_Preferred
This HSS T3 column is a fully endcapped, low-coverage C18 bonded-phase column designed for reversed-phase separation of polar compounds in high aqueous mobile phases. It offers easy scalability and superior repeatability, enabling high-resolution separations for complex samples.
  • HSS (High Strength Silica) technology provides mechanical strength for UPLC pressures and enhanced chromatographic performance.
  • Enhanced silanophilicity increases polar compound retention and selectivity.
  • Low-coverage C18 phase compatible with 100% aqueous mobile phase for polar and non-polar substances.
  • Fully scalable and transferable to UPLC separations.
  • Ultra-low MS bleed for minimal interference in mass spectrometry applications.
  • OBD (Optimum Bed Density) design ensures integrity and stability for quick, high-purity compound isolations.
Catalog No. 50-239-1975