Learn More
Waters Corp Waters XSelect HSS T3 Column, 100Å, 2.5 μm, 3 mm X 50 mm
Supplier: Waters Corp 186006153
The XSelect HSS T3 is a low-coverage C18 phase column compatible with 100% aqueous mobile phase, designed for separating polar and non-polar compounds. These 2.5 μm eXtended Performance (XP) columns provide exceptional separation performance, robustness, and high throughput for HPLC assays, with seamless transferability to UPLC separations.
- Chemistry: C18
- Separation mode: Reversed phase
- Particle substrate: Silica
- pH range: 2 to 8
- Temperature limits: 45°C
- Maximum pressure: 18000 psi (1240 Bar)
- Particle size: 2.5 μm
- Pore size: 100 Å
- Inner diameter: 3 mm
- Length: 50 mm
- System compatibility: UPLC, UHPLC
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.