Learn More
Waters Corp Waters XSelect HSS T3 C18 Column, 2.5 μm, 4.6 mm, 30 mm
Supplier:  Waters Corp 186006156
Encompass_Preferred
The HSS T3 sorbents utilize a trifunctional alkyl C18 bonded phase that is compatible with 100% aqueous mobile phase. This design promotes superior retention of polar compounds and delivers ultra-low MS bleed. These HPLC columns are fully scalable and transferable for UPLC separations.
  • Trifunctional alkyl C18 bonded phase
  • Compatible with 100% aqueous mobile phase
  • Provides ultra-low MS bleed
  • Promotes superior polar compound retention
  • Fully scalable and transferable to UPLC separations
Catalog No. 50-201-561