Learn More
Waters Corp Waters Corporation Column, HSS, 100Å, 2.5 μm, 4.6 mm, 75 mm
Supplier:  Waters Corp 186006158
Encompass_Preferred
The HSS T3 column features a low-coverage C18 phase, compatible with 100% aqueous mobile phase, ideal for developing separations of both polar and non-polar compounds. These columns exhibit ultra-low MS bleed, are fully scalable and transferable to UPLC separations. Packed in ultra-low dispersion hardware, the 2.5 μm XP columns offer exceptional separation performance, robustness, and throughput for HPLC assays, facilitating a seamless transition to UPLC.
  • Low-coverage C18 phase compatible with 100% aqueous mobile phase
  • Ideal for developing polar and non-polar compound separations
  • Exhibits ultra-low MS bleed
  • Fully scalable and transferable to UPLC separations
  • Packed in ultra-low dispersion hardware
  • Delivers exceptional separation performance, robustness, and throughput for HPLC assays
  • Seamless transition path toward UPLC adoption
Catalog No. 50-201-563