Learn More
Waters Corp HPLC Column, XSelect HSS T3, 2.5 μm, 4.6 mm x 50 mm, Method Validation Kit
Supplier:  Waters Corp 186006261
Encompass_Preferred
This HPLC column features XSelect HSS T3 chemistry with a 2.5 μm particle size, 4.6 mm inner diameter, and 50 mm length. Designed for high-speed applications, it is part of a Method Validation Kit that contains three columns from different material batches. The column operates within a pH range of 2 to 8 and has a 100 Å pore size.
  • HSS T3 chemistry with trifunctional C18 ligand type.
  • 2.5 μm particle size; 4.6 mm inner diameter; 50 mm length.
  • 100 Å pore size and 11% carbon load.
  • Endcapped, spherical particles with 230 m²/g medium silanol activity.
  • Unmatched selectivity and flexibility, directly scalable to HPLC and UPLC.
  • Ultra-high pressure compatible, providing lower backpressure.
  • Seamless integration with any LC system for rapid method optimization.
  • Method Validation Kit includes three columns from different material batches.
Catalog No. 50-201-596