Learn More
Waters Corp Waters Corporation XSelect HSS T3 XP Column, 2.1 x 30 mm, 2.5 μm
Supplier:  Waters Corp 176002626
Encompass_Preferred
The XSelect HSS T3 XP Columns are engineered to enhance chromatographic performance by offering increased retention for polar compounds compared to standard C18 phases. These reversed-phase columns are well-suited for use with up to 100% aqueous mobile phases and provide extensive retention capabilities for mixtures of both polar and non-polar compounds when employing standard reversed-phase mobile phases.
  • Increased retention for polar compounds.
  • Compatible with up to 100% aqueous mobile phases.
  • Provides widest retention capability for mixtures of polar and non-polar compounds.
  • Offers exceptional separation performance and robustness.
  • Enables 2-4x improvement in HPLC productivity.
  • Fully compatible with HPLC, UHPLC, and UPLC technology platforms.
  • Designed to withstand higher pressures.
Catalog No. 50-179-285