Learn More
Waters Corp Column, Peptide CSH C18, 2.5 μm, 100 mm
Supplier:  Waters Corp 186006947
Encompass_Preferred
This column features CSH130 C18 based particles with a low, carefully-defined concentration of positive charges, ensuring excellent peak shape for peptides with FA- or TFA-containing mobile phases. Its performance is largely independent of strong ion pairing agents, making it suitable for LC or LC-MS applications. Packed in ultra-low dispersion hardware, it provides exceptional separation performance, robustness, and high throughput for HPLC assays, and offers a smooth transition to UPLC adoption.
  • Excellent peak shape for peptides using FA- or TFA-containing mobile phases due to controlled positive charges.
  • Minimal dependence on strong ion pairing agents, ideal for LC or LC-MS applications.
  • Ultra-low dispersion hardware for exceptional separation performance.
  • Robustness and high throughput for HPLC assays.
  • Seamless transition path toward UPLC adoption.
Catalog No. 50-239-2588