Learn More
Waters Corp Waters Column, CSH C18, 3.5μm, 2.1 mm x 150 mm
Supplier:  Waters Corp 186006952
Encompass_Preferred
Waters XSelect CSH130 C18 3.5 μm particles contain a low and carefully-defined concentration of positive charges. This characteristic results in excellent peak shape for peptides when using either FA- or TFA-containing mobile phases. The column's performance shows minimal dependence on strong ion pairing agents, making it suitable for LC or LC-MS applications.
  • Low and carefully-defined concentration of positive charges for excellent peak shape with peptides.
  • Exhibits little dependence on strong ion pairing agents.
  • Ideal for LC or LC-MS applications.
Catalog No. 50-203-7780