Learn More
Waters Corp Method Validation Kit, XSelect Peptide CSH C18, 130Å, 2.5 μm, 4.6 mm x 100 mm
Supplier:  Waters Corp 186006966
Encompass_Preferred
This method validation kit includes three columns packed with different batches of CSH130 C18 XP sorbent, designed for high-performance peptide separations.
  • Designed for method validation, includes three columns with different sorbent batches.
  • CSH130 C18 particles feature low, precisely defined positive charges for excellent peptide peak shape.
  • Suitable for LC or LC-MS applications, performance independent of strong ion pairing agents.
  • Ultra-low dispersion hardware provides exceptional separation, robustness, and throughput for HPLC assays.
  • Offers a seamless transition path to UPLC adoption.
Catalog No. 50-239-2641