Learn More
Waters Corp Method Validation Kit, C18, 3.5 μm, 2.1 mm x 100 mm, 130 Å
Supplier:  Waters Corp 176003080
Encompass_Preferred
The XSelect Peptide CSH C18 Method Validation Kit is designed for robust peptide separations. It includes three CSH C18 columns from different batches and a Cytochrome C Digestion Standard for benchmarking, complete with a certificate of analysis. CSH™ technology ensures excellent peak shape for peptides in LC or LC-MS applications.
  • Kit contains three CSH130 C18 columns from different material batches.
  • Charged surface hybrid particles facilitate use with various acid mobile phases.
  • Provides superior peptide separations in LC and LC-MS applications.
  • Offers outstanding peak capacities for peptides up to 10,000 Daltons.
  • Ensures excellent mass loading for complex peptide samples.
  • Scalable chemistries from nano flow to lab-scale purification.
  • QC tested for column-to-column reproducibility.
  • Includes Cytochrome C Digestion Standard for benchmarking.
Catalog No. 50-239-1107