Learn More
Waters Corp XSelect Column, C18, 2.5 μm, 2.1 mm × 150 mm, 100 Å
Supplier:  Waters Corp 186008760
Encompass_Preferred
The XSelect Peptide HSS T3 Column is designed for separations requiring silica-based selectivities or increased retention of hydrophilic peptides. It features High Strength Silica (HSS) particle technology, ensuring excellent mechanical stability, long column lifetimes, and high efficiencies for UPLC and UHPLC systems, particularly in LC-MS methods.
  • Designed for separations where silica-based selectivities are desired or increased retention of hydrophilic peptides is required.
  • Each batch undergoes rigorous quality control testing, including gradient separation of a tryptic digest of cytochrome c and chromatographic testing with acidic, basic, and neutral analytes.
  • Ideal for use with low dispersion (UPLC or UHPLC) systems, especially for LC-MS methods.
  • Manufactured in a cGMP, ISO 9001:2000 certified plant, with a final quality control test for packed bed efficiency on each column.
Catalog No. 50-239-1036