Learn More
Waters Corp XSelect Peptide HSS T3 Column, 100 Å Pore, 5 μm Particle
Supplier:  Waters Corp 176004041
Encompass_Preferred
XSelect Peptide HSS T3 columns utilize High Strength Silica (HSS) particle technology, providing increased peptide retentivity and distinct selectivity for peptide separations.
  • Manufacturing in a cGMP, ISO 9001:2000 certified plant ensures narrow tolerances.
  • Each batch chromatographically tested with acidic, basic, and neutral analytes.
  • QC testing with tryptic digest of cytochrome c to pass stringent performance criteria.
  • Final QC test for packed bed efficiency performed on each column; results available.
  • Optional eConnect electronic column tags for identification and tracking.
  • Reliable operation across a broad pH range of 2 to 8.
  • Aids in achieving reproducible chromatography and desired chromatographic resolution.
Catalog No. 50-239-1616