Learn More
Waters Corp XSelect Peptide HSS T3 Column, 100 Å, 2.5 μm, 4.6 x 50 mm
Supplier:  Waters Corp 176004007
Encompass_Preferred
XSelect Peptide HSS T3 Columns utilize High Strength Silica (HSS) particle technology to deliver increased peptide retentivity and different selectivity, suitable for various chromatographic applications.
  • Contains high strength silica (HSS) particle technology.
  • Delivers increased peptide retentivity.
  • Offers different selectivity.
  • Manufactured in a cGMP, ISO 9001:2000 certified plant.
  • Each batch chromatographically tested with acidic, basic, and neutral analytes.
  • Each batch of HSS T3, 100 Å material is QC tested with a gradient separation of a tryptic digest of cytochrome c and passes stringent performance criteria.
  • A final QC test for packed bed efficiency is performed on each column.
  • Recommended operating pH range is 2 to 8.
Catalog No. 50-239-1548