Learn More
Waters Corp HPLC Column, XSelect Peptide HSS T3 C18, 4.6 mm ID, 100 mm, 2.5 μm
Supplier:  Waters Corp 186008763
Encompass_Preferred
Peptide HSS T3 columns are designed for separations where silica-based selectivities are desired or increased retention of hydrophilic peptides is required. This version of the XSelect Peptide HSS T3 column has an inner diameter of 4.6 mm, a length of 100 mm and a particle size of 2.5 μm.
  • Designed for separations requiring silica-based selectivities or increased retention of hydrophilic peptides.
  • Enables reproducible and highly selective chromatographic separations.
  • Utilizes Charged Surface Hybrid (CSH) and High Strength Silica (HSS) particle technologies.
  • Allows for a broad range of selectivities in method development without compromising performance.
Catalog No. 50-239-1619