Learn More
Waters Corp XSelect Peptide HSS T3 Column, 100Å, 5 μm, 4.6 mm x 250 mm
Supplier:  Waters Corp 186008870
Encompass_Preferred
XSelect Peptide HSS T3 Column, 100Å, 5 μm, 4.6 mm X 250 mm columns are designed for separations where silica-based selectivities are desired or when increased retention of hydrophilic peptides are required. Each batch of HSS T3, 100Å material is QC tested with a gradient separation of a tryptic digest of cytochrome c and must pass stringent performance criteria before acceptance for use in these columns.
  • Designed for separations where silica-based selectivities are desired or when increased retention of hydrophilic peptides are required.
  • Each batch of HSS T3, 100Å material is QC tested with a gradient separation of a tryptic digest of cytochrome c and must pass stringent performance criteria before acceptance for use in these columns.
  • Suitable for peptide separations and analysis.
Catalog No. 50-239-2174