Learn More
Waters Corp XSelect Peptide HSS T3 Column, 100 Å, 5 μm, 2.1 x 150 mm
Supplier: Waters Corp 186008787
These XSelect Peptide HSS T3 columns are designed for separations requiring silica-based selectivities or increased retention of hydrophilic peptides. Each batch is QC tested, and a Method Validation Kit (MVK) with three columns is included.
- C18 chemistry, reversed phase, HSS particle technology.
- Silica substrate; spherical, 5 μm particle size, 100 Å pore.
- pH range 2 to 8; up to 45°C; 6000 psi (415 bar) max pressure.
- Endcapped with T3 bonding, medium silanol activity.
- Molecular weight range: 1000 to 15000.
- Peptide QC tested, USP L1, for HPLC systems.
- Dimensions: 2.1 mm ID x 150 mm length; 11% carbon load.
- 3-pack available.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.