Learn More
Waters Corp XSelect Peptide HSS T3 Column, 100Å, 2.5 μm, 4.6 mm × 150 mm
Supplier: Waters Corp 186008784
These columns utilize High Strength Silica (HSS) particle technology, enhancing peptide retentivity and offering distinct selectivity for demanding peptide separations, ensuring high-quality, reproducible chromatographic results.
- High Strength Silica (HSS) particle technology for enhanced peptide retention.
- Offers distinct selectivity.
- Manufactured in a cGMP, ISO 9001:2000 certified facility.
- Rigorous chromatographic testing with acidic, basic, and neutral analytes.
- QC tested using gradient separation of cytochrome c tryptic digest.
- Final QC test for packed bed efficiency on each column.
- Suitable for peptide applications; recommended operating pH 2 to 8.
- Withstands pressures up to 18,000 psi (2.5 μm XP columns).
- Maximum recommended temperature is 45 °C.
- Provides reproducible chromatography and desired resolution.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.