Learn More
Waters Corp XSelect Peptide HSS T3 Column, 100 Å, 5 μm, 2.1 x 100 mm
Supplier: Waters Corp 176004017
This kit includes one Peptide HSS T3 column and one vial of Cytochrome C Digestion Standard. The standard allows immediate benchmarking upon receipt and can be reordered for continued monitoring. Each batch of digestion standard is prepared under strict quality control criteria confirmed on the included certificate of analysis.
- C18 chemistry
- Reversed phase separation mode
- Silica particle substrate
- pH range of 2 to 8
- Temperature limits up to 45°C
- Maximum pressure of 6000 psi (415 bar)
- T3 bonding technology
- 5 μm particle size
- 100 Å pore size
- 2.1 mm inner diameter
- 100 mm length
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.