Learn More
Waters Corp Column, C18, 3.5 μm, 2.1 x 150 mm
Supplier:  Waters Corp 186010636
Encompass_Preferred
These general purpose C18 columns use Charged Surface Hybrid (CSH) technology for superior peak shape and increased loading capacity for basic compounds. They offer excellent separation performance, robustness, and throughput for HPLC assays. MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions, preventing poor peak shape and signal loss.
  • General purpose C18 columns offer alternative selectivity.
  • CSH technology provides superior peak shape and increased loading capacity for basic compounds.
  • Exceptional separation performance, robustness, and throughput for HPLC assays.
  • Seamless transition path toward UPLC adoption.
  • MaxPeak High Performance Surface hardware reduces unwanted analyte/surface interactions.
  • Prevents poor peak shape and signal loss.
Catalog No. 50-288-6202