Learn More
Waters Corp XSelect Column, CSH C18, 3.5 μm, 4.6 x 250 mm
Supplier:  Waters Corp 186010645
Encompass_Preferred
These general purpose C18 columns offer alternative selectivity for HPLC assays. Featuring Charged Surface Hybrid (CSH) technology for superior peak shape and MaxPeak High Performance Surface hardware to reduce analyte interactions, they provide exceptional separation performance and robustness.
  • Provides alternative selectivity compared to other C18 sorbents.
  • Charged Surface Hybrid (CSH) technology enables superior peak shape and increased loading capacity for basic compounds.
  • Exceptional separation performance, robustness, and throughput for HPLC assays.
  • Facilitates a seamless transition path toward UPLC adoption.
  • MaxPeak High Performance Surface hardware reduces unwanted analyte/surface interactions, improving peak shape and signal intensity.
Catalog No. 50-288-6255