Learn More
Waters Corp Column, PFP, 2.5 μm, 2.1 mm x 50 mm
Supplier:  Waters Corp 186010043
Encompass_Preferred
The XSelect Premier HSS PFP Column is a non-endcapped, silica-based column designed for low pH separations, offering alternative selectivity. Its MaxPeak High Performance Surface hardware significantly reduces unwanted analyte/surface interactions, improving peak shape and minimizing signal intensity losses. This HSS Technology provides increased retentivity and broad selectivity.
  • Designed for low pH separations.
  • Features MaxPeak High Performance Surface technology.
  • Reduces analyte/surface interactions and improves peak shape.
  • Provides increased retentivity and broad selectivity.
  • Ensures mechanical stability and improves sensitivity for metal-sensitive analytes.
  • Reduces conditioning and method development time, and improves recovery.
Catalog No. 50-239-1632