Learn More
Waters Corp Column, XSelect Premier HSS T3, 100 Å, 2.5 μm, 2.1 X 100 mm
Supplier:  Waters Corp 186009831
Encompass_Preferred
The XSelect Premier HSS T3 column is a low-coverage C18 phase, compatible with 100% aqueous mobile phase, ideal for separating polar and non-polar compounds. It offers ultra-low MS bleed, scalability to UPLC, and exceptional performance for UHPLC and HPLC assays. MaxPeak High Performance Surface technology reduces unwanted analyte/surface interactions and prevents metal-specific adsorption. Its high-strength silica structure and excellent pH stability handle diverse compounds.
  • Low-coverage C18 phase, 100% aqueous compatible.
  • Ideal for polar and non-polar compound separations.
  • Ultra-low MS bleed.
  • Scalable and transferable to UPLC.
  • MaxPeak High Performance Surface technology reduces unwanted analyte/surface interactions.
  • Excellent reversed-phase retention of polar and non-polar analytes.
  • Excellent pH stability.
Catalog No. NC3419599