Learn More
Waters Corp XSelect Peptide CSH C18 Column, 130 Å, 2.5 μm
Supplier:  Waters Corp 186010715
Encompass_Preferred
The XSelect Premier Peptide CSH C18 Column is optimized for peptide separations, delivering excellent peak shape with FA- or TFA-containing mobile phases. It’s suitable for LC or LC-MS applications and enhances reproducibility for metal-sensitive analytes with an integrated, replaceable 5mm pre-column.
  • Excellent peak shape for peptides using FA- or TFA-containing mobile phases due to carefully-defined positive charges.
  • Quality controlled with cytochrome c digest standards for reliable peptide analysis.
  • Ideal for LC or LC-MS applications with minimal dependence on strong ion pairing agents.
  • Integrated, replaceable 5mm pre-column protects and extends analytical column life without impacting resolution.
Catalog No. 50-239-0684