Learn More
Waters Corp Waters Column, XSelect Premier Peptide CSH C18, 2.5 μm, 2.1 x 150 mm
Supplier:  Waters Corp 186009906
Encompass_Preferred
This column is designed for peptide separations, offering improved peak shape and minimal dependence on strong ion pairing agents, making it suitable for LC or LC-MS applications.
  • Features particles with positive charges for enhanced peptide peak shape.
  • Quality controlled with cytochrome c digest standards for peptide analysis.
  • Recommended for low dispersion (UPLC or UHPLC) systems, especially for LC-MS methods.
  • Increases reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Utilizes Charged Surface Hybrid (CSH) particle technology for balanced interactions.
  • Offers low anion exchange, ideal for protein-related assays.
  • Provides alternate selectivity compared to traditional C18 columns.
  • Maintains excellent pH stability, rated for 1 to 11 pH.
  • Prevents unwanted analyte-surface interactions with MaxPeak technology.
Catalog No. 50-222-4323