Learn More
Waters Corp XSelect Premier Peptide HSS T3 Column, 100 Å, 2.5 μm, 2.1 x 150 mm, VanGuard FIT Guard
Supplier:  Waters Corp 186010726
Encompass_Preferred
Waters Corporation XSelect Premier Peptide HSS T3 columns with MaxPeak High Performance Surfaces are designed for the separation of small, polar peptides, offering greater retentivity and minimizing analyte/surface interactions. This technology enhances analyte recovery, sensitivity, and reproducibility for various bioseparations.
  • Reduced column conditioning and passivation times.
  • Improved sensitivity and peak shapes.
  • Improved recovery.
  • Simpler mobile phases, without complex additives.
  • Time savings in method development.
  • Reduced risk and greater confidence in data and decision making.
  • Mitigates the loss of metal-sensitive analytes such as lipids, organic acids, acidic peptides, new modalities including oligonucleotides, or other compounds containing phosphate or carboxylate functionalities, larger nucleic acids, and plasmids.
Catalog No. 50-239-2846