Learn More
Waters Corp XSelect Premier Peptide HSS T3 Column, 100 Å, 2.5 μm, 2.1 x 150 mm
Supplier:  Waters Corp 186009841
Encompass_Preferred
The XSelect Premier Peptide HSS T3 Column facilitates separations requiring silica-based selectivities or increased retention of hydrophilic peptides. Tested for stringent performance, these 2.1 mm ID columns are ideal for low dispersion UPLC or UHPLC systems, particularly for LC-MS methods. Part of the MaxPeak Premier class, these columns enhance reproducibility, peak shape, and recovery for metal-sensitive analytes. They are 100% water wettable and provide excellent low pH stability.
  • Increased retention of hydrophilic peptides
  • Part of MaxPeak Premier class
  • Prevents unwanted surface-analyte interactions
  • Maximizes selectivity for best performance
  • 100% water wettable, usable with 100% aqueous mobile phase
  • Excellent low pH stability for acidic analytes
  • Solid silica core for easier data reproducibility
  • MaxPeak High Performance Surface Technology protects assays
Catalog No. 50-239-1887