Learn More
Waters Corp XSelect Premier Peptide HSS T3 Column, 100Å, 2.5 μm, 2.1 x 50 mm
Supplier:  Waters Corp 186009839
Encompass_Preferred
This Premier Peptide HSS T3 Column delivers silica-based selectivities and increased retention for hydrophilic peptides. Optimized for low dispersion (UPLC/UHPLC) and LC-MS methods, it enhances reproducibility, peak shape, and recovery for metal-sensitive analytes. Stable HSS particles ensure mechanical and low pH stability, while MaxPeak technology prevents unwanted analyte interactions.
  • Silica-based selectivity and increased retention of hydrophilic peptides.
  • Enhanced reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Batch QC tested for stringent performance criteria.
  • MaxPeak High-Performance Surface technology prevents analyte-surface interactions.
  • Stable HSS particle offers mechanical and low pH stability.
  • 100% water wettable, suitable for aqueous mobile phases and polar analyte retention.
  • Facilitates method development and transfer.
Catalog No. 50-222-4258