Learn More
WELCH MATERIALS INC Xtimate C18, 5µm, 30×250mm
Supplier:  WELCH MATERIALS INC 0161021108
Encompass_Preferred
Prep Column, Xtimate C18, 5µm, 30×250mm, pH range: 1.0-12.5, Pore Size: 120Å, USP listing: L1, Carbon Loading: 14%, Surface Area: 320m2/g, endcapped.
Catalog No. NC2052720
Only null left
Add to Cart
Request a Quote