Learn More
WELCH MATERIALS INC Xtimate CN, 5µm, 120Å, 4.6×250mm
Supplier:  WELCH MATERIALS INC 0010521043
Encompass_Preferred
Analytical Column, Xtimate CN, 5μm, 120Å, 4.6×250mm, pH range: 1.0-12.5, Pore Size: 120Å, USP listing: L10, USP listing: L10, Carbon Loading: 7%, Surface Area: 320m2/g, endcapped.
Catalog No. NC2052231