Learn More
WELCH MATERIALS INC Xtimate Phenyl-Hexyl, 3µm, 120Å, 4.6×250mm
Supplier:  WELCH MATERIALS INC 0010411043
Encompass_Preferred
Analytical Column, Xtimate Phenyl-Hexyl, 3μm, 120Å, 4.6×250mm, pH range: 1.0-12.5, Pore Size: 120Å, USP listing: L11, Carbon Loading: 12%, Surface Area: 320m2/g, endcapped.
Catalog No. NC2052233