Learn More
Waters Corp XTerra MS C18 Sentry Guard Cartridge, 125Å, 3.5 µm, 3 mm X 20 mm, 2/pk
Supplier:  Waters Corp 186000640
Encompass_Preferred
XTerra MS C18 columns are general purpose, hybrid-based, reversed-phase C18 columns that are designed to be compatible with mass spectrometry applications. These first generation columns provide superior pH stability compared to a silica-based column.
Catalog No. 50-819-072