Learn More
Waters Corp XTerra MS C18 Column, 125Å, 2.5 μm, 2.1 mm x 50 mm
Supplier:  Waters Corp 186000594RF
Encompass_Preferred
This XTerra MS C18 Column is designed for high-performance liquid chromatography and features an eConnect Column Tag for digital integration and enhanced laboratory efficiency.
  • Features an eConnect tag for digital integration.
  • Stores unique product identifying information, automatically imported by Empower CDS.
  • NFC enabled for web access and smart device support.
  • Pre-equipped tag simplifies setup and reduces manual data entry.
  • Tamper-proof design ensures compliance and auto-detection by Alliance iS.
  • Tag design prevents interference in temperature-sensitive LC methods.
  • Identical packing to untagged versions ensures consistent quality.
Catalog No. 50-239-2508