Learn More
Waters Corp Waters Corporation Column, MS C18, 3.5 μm, 2.1 × 50 mm, 125 Å
Supplier:  Waters Corp 186000400
Encompass_Preferred
XTerra MS C18 columns are general-purpose, hybrid-based, reversed-phase C18 columns designed for compatibility with mass spectrometry applications. These first-generation columns offer superior pH stability compared to silica-based columns, providing great separation efficiency and improved pH stability.
  • Designed for mass spectrometry applications, featuring crisp peak forms, high sensitivity, and large peak capacities.
  • Offers long column life over a wide pH range (1 to 12) due to tri-functional bonding chemistry.
  • Provides maximum throughput with exceptional resolution and minimal column bleed.
  • Manufactured using ultrapure reagents, ensuring controlled chemical composition and purity of the hybrid particle.
  • Each column is individually examined to guarantee high-quality laboratory equipment.
  • Withstands a pH range of 1 to 12 and a maximum pressure of 6000 psi (415 Bar).
Catalog No. 50-819-146