Learn More
Waters Corp Waters XTerra MS C8 Column, 3.5 μm, 4.6 × 150 mm, 125 Å
Supplier:  Waters Corp 186000441
Encompass_Preferred
The XTerra MS C8 Column is a general-purpose, hybrid-based, reversed-phase C8 column for mass spectrometry (MS) and liquid chromatography-mass spectrometry (LC-MS) applications. It offers superior pH stability and efficient separations compared to silica-based columns.
  • C8 chemistry for reversed-phase separation
  • Hybrid particle substrate; spherical particle shape
  • pH range of 1 to 12; maximum pressure of 6000 psi (415 Bar)
  • Endcapped with MS C8 bonding technology
  • Low silanol activity
  • 3.5 μm particle size and 125 Å pore size
  • 4.6 mm inner diameter and 150 mm length
  • 12% carbon load
  • Compatible with HPLC systems and diverse mobile phases
  • Suitable for analysis of peptides, proteins, and small molecules
  • Provided in a single-pack format
Catalog No. 50-818-713