Learn More
Waters Corp XTerra Column, Phenyl, 3.5 μm, 125 Å, 2.1 mm x 150 mm
Supplier:  Waters Corp 186001181
Encompass_Preferred
XTerra Phenyl columns are hybrid-based, reversed-phase columns that incorporate phenyl ligand functionality. These first-generation columns offer superior pH stability compared to silica-based columns. This column is meticulously designed for precise separation of diverse polar and non-polar compounds. Crafted from 125Å porous silica, modified with phenyl groups, it offers exceptional retention capabilities. It is compatible with various HPLC systems and detectors, allowing for seamless integration into analytical workflows.
  • Exceptional retention capabilities.
  • Compatibility across various HPLC systems and detectors.
  • Precision engineering ensures stability and reproducible performance.
  • Reduced dead volume and extended column lifetime.
  • Column lifetime can be extended with a compatible VanGuard Cartridge.
Catalog No. 50-780-612