Learn More
Waters Corp WATERS CORPORATION XTerra Phenyl Column, 125 Å, 5 μm, 3.9 mm x 150 mm
Supplier:  Waters Corp 186001184
Encompass_Preferred
These hybrid-based, reversed-phase columns with phenyl ligand functionality offer superior pH stability and complementary selectivity for aromatic compounds. They provide excellent surface coverage, robust pH stability, consistent retention times, and good, reproducible peak shapes. Made with Hybrid Particle Technology, they ensure enhanced separation efficiency and improved pH stability. Each is produced with ultrapure chemicals and tested, with Certificates of Batch Analysis and Column Efficiency.
  • Hybrid-based, reversed-phase with phenyl ligand functionality
  • Superior and robust pH stability over silica-based columns
  • Complementary selectivity for aromatic compounds
  • Excellent surface coverage
  • Consistent, reproducible retention times, good peak shapes
  • Di-functional bonding enhances low pH hydrolytic stability
  • Available in various porosity and particle sizes
Catalog No. 50-780-614