Learn More
Waters Corp Waters Corporation Phenyl Column, 5 μm, 4.6 mm, 150 mm, 125 Å
Supplier:  Waters Corp 186001146
Encompass_Preferred
Phenyl columns are hybrid-based, reversed-phase columns with phenyl ligand functionality. These first-generation hybrid particle stationary phases combine silica's efficiency with polymer stability, offering superior pH stability (1 to 12 pH range) compared to silica-based columns. They deliver excellent peak shapes and complementary selectivity.
  • Hybrid particle type (methylene bridged, difunctional bonding)
  • Phenyl ligand functionality
  • Spherical particle shape
  • Low silanol activity
  • 125 Å pore size
  • 12.0% carbon load
  • High-efficiency separations
  • High mechanical strength and stability
  • Excellent peak shapes for compounds, including bases
  • Mass Spectrometry (MS) compatible
  • Easy scale-up for chromatography
  • Increased mass loading at high pH (up to 60x)
Catalog No. 50-780-578