Learn More
Waters Corp Waters Corporation Phenyl Column, 5 μm, 125 Å, 4.6 mm, 100 mm
Supplier:  Waters Corp 186001145
Encompass_Preferred
XTerra Phenyl columns are hybrid-based, reversed-phase columns that incorporate phenyl ligand functionality. These first generation columns provide superior pH stability compared to a silica-based column.
  • Incorporates phenyl ligand functionality.
  • Provides superior pH stability compared to a silica-based column.
  • Offers a complementary selectivity to other phases.
  • Delivers excellent peak shapes for all compounds.
  • Has a pH range of 1–12.
  • Ideally suited for Mass Spec applications.
Catalog No. 50-780-577