Learn More
Waters Corp Waters Corporation Column, MS C18, 10 μm, 125 Å, 50 x 50 mm
Supplier:  Waters Corp 186002279
Encompass_Preferred
This XTerra MS C18 preparative column is designed for high-performance liquid chromatography, optimized for Mass Spectrometry applications. It combines silica particle efficiency and polymer particle pH stability, offering robust performance, excellent peak shapes for bases, and a broad operational pH range.
  • Compatible with mass spectrometry.
  • Delivers sharp peaks, good sensitivity, and high peak capacities.
  • Trifunctional bonding ensures long lifetimes and a wide pH range.
  • Provides high throughput, excellent resolution, and ultra-low bleed.
  • Combines silica efficiency with polymer pH stability for rugged performance.
  • Features a pH range of 1–12.
  • Facilitates easy scale-up from analytical to preparative chromatography.
  • High pH operation improves peak shapes for bases and increases mass loading.
Catalog No. 50-781-055
Add to Cart
Request a Quote