Learn More
Waters Corp Waters Column, 30 mm x 250 mm, 10 μm, 125 Å
Supplier:  Waters Corp 186002272
Encompass_Preferred
This preparative column is designed for Mass Spectrometry applications, providing sharp peaks, good sensitivity, and large peak capacities. It incorporates trifunctional bonding chemistries, which ensures long lifetimes across a wide pH range and enables high throughput with excellent resolution and ultra-low bleed. The column is ideally suited for Mass Spec and allows for easy scale-up from analytical to preparative chromatography.
  • Designed for Mass Spectrometry applications
  • Offers sharp peaks, good sensitivity, and large peak capacities
  • Features trifunctional bonding chemistries
  • Provides long lifetimes across a wide pH range of 1–12
  • Enables high throughput with excellent resolution and ultra-low bleed
  • Allows for easy scale-up from analytical to preparative chromatography
Catalog No. 50-781-048
Add to Cart
Request a Quote