Learn More
Waters Corp Waters Corporation XTerra Shield RP18 Prep Column, 10 μm, 7.8 mm × 300 mm
Supplier:  Waters Corp 186001166
Encompass_Preferred
The XTerra Shield RP18 Prep Column is a hybrid-based, reversed-phase C18 column featuring patented embedded polar group technology. This design improves peak shape performance for highly basic compounds and offers superior pH stability (pH range of 2-12) compared to traditional silica-based columns.
  • Hybrid particle substrate for high pH stability
  • 10 μm particle size, 125 Å pore size
  • 7.8 mm inner diameter, 300 mm length
  • Embedded polar group technology improves peak shape of basic compounds
  • Organic/inorganic Hybrid Particle Technology (HPT) for enhanced mechanical strength
  • Broad operating pH range of 2 to 12, temperature limit of 60°C
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped with Shield RP18 bonding technology
  • Low silanol activity, spherical particle shape
  • Suitable for HPLC Preparative, LC/MS, and LC (USP Classification L1)
Catalog No. 50-780-598