Learn More
Waters Corp WATERS RP18 Prep Column, 5 μm, 30 mm X 100 mm, 125 Å
Supplier:  Waters Corp 186001944
Encompass_Preferred
This hybrid-based, reversed-phase C18 column utilizes embedded polar group technology to enhance peak shape for highly basic compounds. It offers superior pH stability compared to silica-based columns and is constructed from a high purity methyl hybrid material. The column is built to ISO 9001 standards.
  • Overcomes hydrolytic instability of silica-based reversed-phase packing materials at high pH.
  • Features organic/inorganic Hybrid Particle Technology (HPT).
  • Maintains high separation efficiency for extremely basic substances.
  • Provides mechanical strength, spherical shape, purity, and optimized pore dimensions.
  • Offers tailored surface area and particle size.
Catalog No. 50-780-835
Add to Cart
Request a Quote