Learn More
Waters Corp WATERS CORPORATION XTerra Shield RP8 Guard Cartridge, 10 μm, 10 x 10 mm
Supplier:  Waters Corp 186001009
Encompass_Preferred
This XTerra HPLC Column is a Shield RP8 type guard cartridge designed with a 10 μm particle size and dimensions of 10 x 10 mm. It facilitates easy scale-up from analytical to preparative chromatography. The column's ability to operate at high pH delivers excellent peak shapes for bases and offers significantly increased mass loading compared to conventional silica-based materials.
  • Easy scale-up from analytical to preparative chromatography.
  • Operates at high pH, providing excellent peak shapes for bases.
  • Increased mass loading up to 60x compared to conventional silica-based materials.
  • 10 x 10 mm Guard Cartridge dimension.
  • 10 μm particle size.
  • Shield RP8 column type.
Catalog No. 50-780-533