Learn More
Waters Corp OBD Column, Shield RP8, 10 μm, 19 x 150 mm
Supplier:  Waters Corp 186002258
Encompass_Preferred
These hybrid-based, reversed-phase C8 preparative columns utilize a first-generation hybrid particle (silica co-polymer) stationary phase. This combines silica's high efficiency with a polymer's expanded pH range, offering ruggedness, high mechanical strength, and an embedded polar group for enhanced peak shape with basic compounds.
  • Embedded polar group technology delivers optimal peak shape and unique selectivity, ensuring excellent water wettability.
  • Offers superior pH stability with a wider pH range (2 to 12) compared to silica-based columns, allowing greater selectivity choices and improved peak resolution.
  • Provides high mechanical strength and efficiency due to hybrid particle technology.
  • Achieves up to 60x increased mass loading compared to conventional silica-based materials.
  • Facilitates easy scale-up from analytical to preparative chromatography.
Catalog No. 50-781-034