Learn More
Waters Corp Waters Corporation XTerra Shield RP8 Column, 3.5 μm, 3.9 mm × 100 mm, 125 Å
Supplier:  Waters Corp 186000429
Encompass_Preferred
XTerra Shield RP8 columns are hybrid-based, reversed-phase C8 columns offering superior pH stability and improved peak shape for basic compounds. They use proprietary organic/inorganic hybrid particle technology (HPT) to combat hydrolytic instability at high pH.
  • General purpose, hybrid-based, reversed-phase C8 column.
  • Improves peak shape for highly basic compounds with embedded polar technology.
  • Offers superior pH stability vs. silica-based columns, utilizing HPT for high pH hydrolytic stability.
  • Constructed with high purity methyl hybrid material.
  • Produced to ISO 9001 guidelines; independently evaluated; batch analysis and column efficiency certificates.
  • Features mechanical strength, spherical shape, purity, and customizable particle size.
  • Excellent separation efficiency for basic chemicals.
  • Broad operating pH range: 2 to 12.
Catalog No. 50-780-248