Learn More
Waters Corp Waters XTerra Shield RP8 Column, C8, 3.5 μm, 4.6 mm ID, 250 mm
Supplier:  Waters Corp 186001473
Encompass_Preferred
XTerra RP8 columns are general-purpose, hybrid-based, reversed-phase C8 columns that utilize patented embedded polar group technology for improved peak shape of highly basic compounds and superior pH stability.
  • Utilizes patented Hybrid Particle Technology for distributed hydrophobicity, enabling high speed, high temperature, and high pH operation.
  • Incorporates patented embedded polar group technology for improved peak shape with highly basic compounds and superior pH stability.
  • Features 33% fewer residual silanols after end-capping and bonding, ensuring sharp, high-efficiency peaks.
  • Manufactured with ultrapure reagents and individually tested for consistent quality.
  • Operable at pressures up to 6000 psi (415 bar), with a pH range of 2 to 12, and a recommended temperature of 20°C to 60°C.
Catalog No. 50-780-702