Learn More
Waters Corp XTerra Shield RP8 Column, 125Å, 5 µm, 3.9 mm X 150 mm, 1/pk
Supplier:  Waters Corp 186000481
Encompass_Preferred
XTerra RP8 columns are general purpose, hybrid-based, reversed-phase C8 columns that incorporate Waters patented embedded polar group technology to improve peak shape performance for highly basic compounds. These first generation columns provide superior pH stability compared to a silica-based column.
Catalog No. 50-780-298