Learn More
Waters Corp XTerra Shield RP18 Column, 125 Å, 5 μm, 4.6 mm X 250 mm
Supplier:  Waters Corp 186000496RF
Encompass_Preferred
The XTerra Shield RP18 Column is for analytical applications, featuring embedded polar group technology for excellent peak shape of basic compounds. This C18 column offers superior pH stability (pH 2 to 12) and high efficiency. Its hybrid construction ensures durability, and it includes an eConnect tag for tracking use history.
  • Embedded polar group technology for basic compounds.
  • Reversed-phase C18 chemistry.
  • Hybrid construction for durability.
  • Accurate analyte separation.
  • Exceptional resolution.
  • Superior pH stability (pH 2 to 12).
  • eConnect tag for tracking.
  • High efficiency.
  • Suited for pharma, environmental, food & beverage, clinical, and industrial analysis.
Catalog No. 50-239-1431