Learn More
Waters Corp XTerra Shield RP18 OBD Prep Column, 125Å, 5 μm, 10 mm x 150 mm
Supplier:  Waters Corp 186008143
Encompass_Preferred
These preparative C18 columns are hybrid-based, reversed-phase, incorporating embedded polar group technology for improved peak shape performance with highly basic compounds. They offer superior pH stability compared to silica-based columns by utilizing distinctive organic/inorganic Hybrid Particle Technology (HPT).
  • Incorporates patented embedded polar group technology to improve peak shape performance for highly basic compounds.
  • Provides superior pH stability compared to silica-based columns, overcoming hydrolytic instability at high pH.
  • Utilizes organic/inorganic Hybrid Particle Technology (HPT) for benefits such as pore size tailoring, purity, and enhanced mechanical strength.
  • Maintains high separation efficiency and optimal shape performance for extremely basic compounds.
  • Constructed in accordance with ISO 9001 standards.
Catalog No. 50-239-1886