Learn More
Waters Corp XTerra Shield RP18 OBD Prep Column, 125Å, 10 μm, 10 mm, 250 mm
Supplier:  Waters Corp 186008135
Encompass_Preferred
XTerra Preparative RP18 columns are hybrid-based, reversed-phase C18 columns that utilize embedded polar group technology. These columns are designed to improve peak shape performance for highly basic compounds and offer superior pH stability compared to silica-based columns.
  • Hybrid-based, reversed-phase C18 columns
  • Incorporates embedded polar group technology
  • Improves peak shape performance for highly basic compounds
  • Provides superior pH stability compared to silica-based columns
  • pH range of 2 to 12
  • Can sustain up to a maximum pressure of 6000 psi (415 bar)
Catalog No. 50-239-2364